2,3-Dimethylnaphthalene, 97%
2,3-Dimethylnaphthalene, 97%
2,3-Dimethylnaphthalene, 97%
Thermo Scientific Chemicals

2,3-Dimethylnaphthalene, 97%

CAS: 581-40-8 | C12H12 | 156.228 g/mol
製品番号(カタログ番号) L08386.03
または、製品番号L08386-03
価格(JPY)
-
数量:
1 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS581-40-8
IUPAC Name2,3-dimethylnaphthalene
Molecular FormulaC12H12
InChI KeyWWGUMAYGTYQSGA-UHFFFAOYSA-N
SMILESCC1=C(C)C=C2C=CC=CC2=C1
さらに表示
Appearance (Color)White to pale cream to black
FormCrystals or powder or crystalline powder or flakes
Assay (GC)≥96.0%
Identification (FTIR)Conforms
Melting Point (clear melt)100.0-109.0?C
It is used in the identification of individual aromatic hydrocarbons in kerosene fraction (b.p. 150-250°).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the identification of individual aromatic hydrocarbons in kerosene fraction (b.p. 150-250°).

Solubility
Solubility in toluene (almost transparency). Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

General References:

  1. Deperasińska I, et al. Single dibenzoterrylene molecules in naphthalene and 2,3-dimethylnaphthalene crystals: vibronic spectra.PCCP.,2011,13(5), 1872-1878.
  2. J.F Messerly, et al. Condensed-phase heat capacities and derived thermodynamic properties for 1,4-dimethylbenzene, 1,2-diphenylethane, and 2,3-dimethylnaphthalene.J. Chem. Thermodyn.,1988,20(4), 485-501.