(1-Hexyl)trimethylammonium bromide, 98%
(1-Hexyl)trimethylammonium bromide, 98%
(1-Hexyl)trimethylammonium bromide, 98%
(1-Hexyl)trimethylammonium bromide, 98%
Thermo Scientific Chemicals

(1-Hexyl)trimethylammonium bromide, 98%

CAS: 2650-53-5 | C9H22BrN | 224.186 g/mol
製品番号(カタログ番号) L08594.14
または、製品番号L08594-14
価格(JPY)
-
見積もりを依頼する
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS2650-53-5
IUPAC Namehexyltrimethylazanium bromide
Molecular FormulaC9H22BrN
InChI KeyJYVPKRHOTGQJSE-UHFFFAOYSA-M
SMILES[Br-].CCCCCC[N+](C)(C)C
さらに表示
Appearance (Color)White
Water Content (Karl Fischer Titration)<2.5%
FormCrystals or powder or crystalline powder
Assay (Titration ex Bromide)≥97.5 to ≤102.5%
Melting Point (clear melt)182-187?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only