Bis(2-oxo-3-oxazolidinyl)phosphinic chloride, 97%
Bis(2-oxo-3-oxazolidinyl)phosphinic chloride, 97%
Bis(2-oxo-3-oxazolidinyl)phosphinic chloride, 97%
Thermo Scientific Chemicals

Bis(2-oxo-3-oxazolidinyl)phosphinic chloride, 97%

CAS: 68641-49-6 | C6H8ClN2O5P | 254.563 g/mol
製品番号(カタログ番号) L08775.14
または、製品番号L08775-14
価格(JPY)
-
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS68641-49-6
IUPAC Namebis(2-oxo-1,3-oxazolidin-3-yl)phosphinoyl chloride
Molecular FormulaC6H8ClN2O5P
InChI KeyKLDLRDSRCMJKGM-UHFFFAOYSA-N
SMILESClP(=O)(N1CCOC1=O)N1CCOC1=O
さらに表示
Assay (Titration ex Chloride)≥96.0 to ≤104.0%
FormCrystals or powder or crystalline powder or lumps
Identification (FTIR)Conforms
Appearance (Color)White to pale cream to pale yellow
Bis(2-oxo-3-oxazolidinyl)phosphinic chloride was used in the preparation of hexadepsipeptide. Also used as reagent is used for activating the carboxylic group, synthesis of amides, esters and peptides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bis(2-oxo-3-oxazolidinyl)phosphinic chloride was used in the preparation of hexadepsipeptide. Also used as reagent is used for activating the carboxylic group, synthesis of amides, esters and peptides.

Solubility
Hydrolyzes with water.

Notes
Moisture Sensitive, store away from moisture. Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only

General References:

  1. T Miyazawa; T Donkai; T Yamada; S Kuwata. Effect of copper(II) chloride on suppression of racemization in peptide synthesis by the mixed anhydride and related methods. International Journal of Peptide and Protein Reseach. 1992, 40 (1), 49-53.
  2. H T Le; J F Gallard; M Mayer; E Guittet; R Michelot. Use of BOP-Cl in the presence of Boc-amino monothioacids for the thioacylation of imino acid residues. Bioorganic & Medicinal Chemistry. 1996, 4(12), 2201-2209.
  3. Coupling reagent for which the rates of reaction with various nucleophiles are sufficiently different to permit a genuine one-step coupling of carboxylic acids with amines: Synthesis, 547 (1980). Particularly suitable for coupling N-alkyl amino acids: J. Org. Chem., 51, 3350 (1986). For discussion of strategies for one-step coupling in synthesis of amides, see: Synthesis, 413 (1984), and of the scope and limitations of the reagent in peptide coupling, see: Int. J. Pept. Prot. Res., 29, 574 (1987); J. Org. Chem., 55, 2895 (1990). See also Appendix 6.
  4. For use in the direct selective 5'-acylation of nucleosides by carboxylic acids, (without 3'-protection), see: Tetrahedron, 44, 229 (1988). For high yield dehydration of carboxylic acids to anhydrides, see: Synthesis, 616 (1981). Also used in macrolide cyclization: J. Am. Chem. Soc., 104, 6818 (1982), and in formation of acyclic esters: Synth. Commun., 14, 515 (1984).
  5. The mixed anhydride formed with carboxylic acids adds to imines to give ß-lactams: Synthesis, 63 (1982):
  6. These are also formed in high yield by cyclization of ß-amino acids: Bull. Korean Chem. Soc., 12, 457 (1991).