cis-4-Cyclohexene-1,2-dicarboxylic acid, 98%
cis-4-Cyclohexene-1,2-dicarboxylic acid, 98%
cis-4-Cyclohexene-1,2-dicarboxylic acid, 98%
cis-4-Cyclohexene-1,2-dicarboxylic acid, 98%
Thermo Scientific Chemicals

cis-4-Cyclohexene-1,2-dicarboxylic acid, 98%

CAS: 2305-26-2 | C8H10O4 | 170.164 g/mol
製品番号(カタログ番号) L09392.22
または、製品番号L09392-22
価格(JPY)
-
見積もりを依頼する
数量:
100 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS2305-26-2
IUPAC Namecyclohex-4-ene-1,2-dicarboxylic acid
Molecular FormulaC8H10O4
InChI KeyILUAAIDVFMVTAU-UHFFFAOYNA-N
SMILESOC(=O)C1CC=CCC1C(O)=O
さらに表示
Assay (Silylated GC)≥97.5% (non-U.S. specification)
FormCrystalline powder
Appearance (Color)White
Assay from Supplier's CofA≥97.5% (U.S. specification)
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5% (non-U.S. specification)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only