Cyclopropylacetic acid, 97%
Cyclopropylacetic acid, 97%
Cyclopropylacetic acid, 97%
Cyclopropylacetic acid, 97%
Thermo Scientific Chemicals

Cyclopropylacetic acid, 97%

CAS: 5239-82-7 | C5H8O22 | 100.12 g/mol
製品番号(カタログ番号) L09416.06
または、製品番号L09416-06
価格(JPY)
-
見積もりを依頼する
数量:
5 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS5239-82-7
IUPAC Name2-cyclopropylacetic acid
Molecular FormulaC5H8O22
InChI KeyBXXOHRWKRXBZTG-UHFFFAOYSA-N
SMILESCN1CCN(C)B1N=S=NB1N(C)CCN1C
さらに表示
Appearance (Color)Clear colorless
Assay (Aqueous acid-base Titration)≥96.0 to ≤104.0%
FormLiquid or viscous liquid
Refractive Index1.4325-1.4385 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only