(S)-(-)-2-Chloropropionic acid, 98%
(S)-(-)-2-Chloropropionic acid, 98%
(S)-(-)-2-Chloropropionic acid, 98%
(S)-(-)-2-Chloropropionic acid, 98%
Thermo Scientific Chemicals

(S)-(-)-2-Chloropropionic acid, 98%

CAS: 29617-66-1 | C3H4ClO2 | 107.51 g/mol
製品番号(カタログ番号) L09427.14
または、製品番号L09427-14
価格(JPY)
-
見積もりを依頼する
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS29617-66-1
Appearance (Color)Clear colorless to pale yellow to yellow to brown
FormLiquid or viscous liquid
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
Assay (GC)≥97.5%
Identification (FTIR)Conforms
さらに表示
Through the stereo displacement of (S)-(-)-2-Chloropropionic acid PPAR Agonist can be synthesized via asymmetric hydrogenation of a cinnamic acid derivative.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Through the stereo displacement of (S)-(-)-2-Chloropropionic acid PPAR Agonist can be synthesized via asymmetric hydrogenation of a cinnamic acid derivative.

Solubility
Fully miscible in water.

Notes
Store in cool dry condition. Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Ioannis N. Houpis.; Lawrence E. Patterson.; Charles A. Alt.; John R. Rizzo.; Tony Y. Zhang.; Michael Haurez. Synthesis of PPAR Agonist via Asymmetric Hydrogenation of a Cinnamic Acid Derivative and Stereospecific Displacement of (S)-2-Chloropropionic Acid.Org. Lett. 2005, 7 (10),1947-1950 .
  2. Atsushi Kurata.; Tatsuo Kurihara.; Harumi Kamachi.; Nobuyoshi Esaki. Asymmetric reduction of 2-chloroacrylic acid to (S)-2-chloropropionic acid by a novel reductase from Burkholderia sp. WSTetrahedron: Asymmetry. 2004, 15 (18),2837-2839 .