1,4-Phenylene diisothiocyanate, 98%
1,4-Phenylene diisothiocyanate, 98%
1,4-Phenylene diisothiocyanate, 98%
Thermo Scientific Chemicals

1,4-Phenylene diisothiocyanate, 98%

CAS: 4044-65-9 | C8H4N2S2 | 192.254 g/mol
製品番号(カタログ番号) L09756.14
または、製品番号L09756-14
価格(JPY)
-
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS4044-65-9
IUPAC Name1,4-diisothiocyanatobenzene
Molecular FormulaC8H4N2S2
InChI KeyOMWQUXGVXQELIX-UHFFFAOYSA-N
SMILESS=C=NC1=CC=C(C=C1)N=C=S
さらに表示
Melting Point (clear melt)127.0-134.0°C
Appearance (Color)White to cream to yellow to brown to gray
Assay (GC)≥97.5% (non-U.S. specification)
Identification (FTIR)Conforms (non-U.S. specification)
Assay from Supplier's CofA≥97.5% (U.S. specification)
さらに表示
It is used in the treatment of hookworms.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the treatment of hookworms.

Solubility
Hydrolyzes in water.

Notes
Store at room temperature. Incompatible with strong oxidizing agents.
RUO – Research Use Only
Thomas Deutschmann.; Helmut Ritter. Polymeric mesoions, 3. Synthesis of polymeric mesoionic 1,3-diazinium-olates via polymer analogous modification of a polythiourea from m-phenylenediamine and p-phenylene diisothiocyanate.Macromol. Rapid Commun. 1996, 17 (10),723-730 .