Thermo Scientific Chemicals

n-Hexyl acrylate, 95%, stab. with hydroquinone, Thermo Scientific Chemicals

Catalog number: L09905.14
25 g, Each
Thermo Scientific Chemicals

n-Hexyl acrylate, 95%, stab. with hydroquinone, Thermo Scientific Chemicals

Catalog number: L09905.14
25 g, Each
Quantity
Catalog number: L09905.14
also known as L09905-14
Price (USD)

Chemical Identifiers

CAS
2499-95-8
IUPAC Name
hexyl prop-2-enoate
Molecular Formula
C9H16O2
InChI Key
LNMQRPPRQDGUDR-UHFFFAOYSA-N
SMILES
CCCCCCOC(=O)C=C
Solution Test
10% in diethyl ether: clear
Form
Liquid
Refractive Index
1.4225-1.4295 @ 20?C
Appearance (Color)
Clear colorless
Assay (GC)
>94.0%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text