Benzofuroxan, 98%
Benzofuroxan, 98%
Benzofuroxan, 98%
Thermo Scientific Chemicals

Benzofuroxan, 98%

CAS: 480-96-6 | C6H4N2O2 | 136.11 g/mol
製品番号(カタログ番号) L10144.09
または、製品番号L10144-09
価格(JPY)
-
数量:
10 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS480-96-6
IUPAC Name2,1,3-benzoxadiazol-1-ium-1-olate
Molecular FormulaC6H4N2O2
InChI KeyOKEAMBAZBICIFP-UHFFFAOYSA-N
SMILES[O-][N+]1=C2C=CC=CC2=NO1
さらに表示
Appearance (Color)Yellow to brown
FormPowder and/or lumpy solid
Melting Point (clear melt)65-71?C
Assay (GC)≥97.5%
Benzofuroxan is used as an oxidant for the preparation of 2-arylsubstituted benzimidazoles, quinoxalines and 1,4-bis(alkylamino)-9,10-anthracenediones. It is also sued in the synthesis of 1,2-dinitrosobenzene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzofuroxan is used as an oxidant for the preparation of 2-arylsubstituted benzimidazoles, quinoxalines and 1,4-bis(alkylamino)-9,10-anthracenediones. It is also sued in the synthesis of 1,2-dinitrosobenzene.

Solubility
Soluble in methanol. Insoluble in water.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Ehlers, D.; Klapötke, T. M.; Pflüger, C. Investigations of the Vicarious C-Aminations of 5,7-Dinitrobenzotriazole and 4,6-Dinitrobenzotriazol-3-ium-1-oxide and Their Energetic Properties. Chem. Eur. J. 2015, 21 (45), 16073-16082.
  2. Liu, Z.; He, J.; Zhuang, H.; Li, H.; Li, N.; Chen, D.; Xu, Q.; Lu, J.; Zhang, K.; Wang, L. Effect of single atom substitution in benzochalcogendiazole acceptors on the performance of ternary memory devices. J. Mater. Chem. C 2015, 3 (35), 9145-9153.