N-(2-Hydroxyphenyl)thiourea, 97%
N-(2-Hydroxyphenyl)thiourea, 97%
N-(2-Hydroxyphenyl)thiourea, 97%
Thermo Scientific Chemicals

N-(2-Hydroxyphenyl)thiourea, 97%

CAS: 1520-26-9 | C7H8N2OS | 168.214 g/mol
製品番号(カタログ番号) L10519.06
または、製品番号L10519-06
価格(JPY)
-
数量:
5 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS1520-26-9
IUPAC Name(2-hydroxyphenyl)thiourea
Molecular FormulaC7H8N2OS
InChI KeyVYBCFZXLXJUFPM-UHFFFAOYSA-N
SMILESNC(=S)NC1=CC=CC=C1O
さらに表示
Appearance (Color)White to cream to brown
FormCrystals or powder or crystalline powder
Assay (HPLC)≥96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only