1,2-O-Isopropylidene-alpha-D-glucofuranose
1,2-O-Isopropylidene-alpha-D-glucofuranose
1,2-O-Isopropylidene-alpha-D-glucofuranose
Thermo Scientific Chemicals

1,2-O-Isopropylidene-alpha-D-glucofuranose

CAS: 18549-40-1 | C9H16O6 | 220.221 g/mol
製品番号(カタログ番号) L10688.06
または、製品番号L10688-06
価格(JPY)
-
数量:
5 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS18549-40-1
IUPAC Name1-{6-hydroxy-2,2-dimethyl-tetrahydro-2H-furo[2,3-d][1,3]dioxol-5-yl}ethane-1,2-diol
Molecular FormulaC9H16O6
InChI KeyBGGCXQKYCBBHAH-UHFFFAOYNA-N
SMILESCC1(C)OC2OC(C(O)CO)C(O)C2O1
さらに表示
Appearance (Color)White to cream
FormPowder or crystalline powder
Water Content (Karl Fischer Titration)<0.5%
Melting point158-163°C
Optical Rotation-10.0° to -12.0° (c=1, water)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only