2-Indanylacetic acid, 99%
2-Indanylacetic acid, 99%
2-Indanylacetic acid, 99%
2-Indanylacetic acid, 99%
Thermo Scientific Chemicals

2-Indanylacetic acid, 99%

CAS: 37868-26-1 | C11H12O2 | 176.22 g/mol
製品番号(カタログ番号) L10700.06
または、製品番号L10700-06
価格(JPY)
-
見積もりを依頼する
数量:
5 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS37868-26-1
Assay (Aqueous acid-base Titration)≥98.5 to ≤101.5%
FormCrystals or powder or crystalline powder or fused solid
Assay (Silylated GC)≥98.5%
Appearance (Color)White to cream to brown
Melting Point (clear melt)88.0-95.0?C
2-Indanylacetic acid is the starting material in the synthesis of pentacylic core of (+)-Salvileucalin.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Indanylacetic acid is the starting material in the synthesis of pentacylic core of (+)-Salvileucalin.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only
Douglass F. Taber,; Craig M. Paquette.Synthesis of the Pentacylic Core of (+)-Salvileucalin B . J. Org. Chem.,. 2014, 79 (8), 3410-3413.