Thermo Scientific Chemicals

Benzyl methacrylate, 98%, stab. with ca 50-100ppm 4-methoxyphenol, Thermo Scientific Chemicals

Catalog number: L10840.30
250 g, Each
Thermo Scientific Chemicals

Benzyl methacrylate, 98%, stab. with ca 50-100ppm 4-methoxyphenol, Thermo Scientific Chemicals

Catalog number: L10840.30
250 g, Each
Quantity
Catalog number: L10840.30
also known as L10840-30
Price (USD)
Price: 72.20
Online price: 61.65
Your price:
Quantity
-

Chemical Identifiers

CAS
2495-37-6
IUPAC Name
benzyl 2-methylprop-2-enoate
Molecular Formula
C11H12O2
InChI Key
AOJOEFVRHOZDFN-UHFFFAOYSA-N
SMILES
CC(=C)C(=O)OCC1=CC=CC=C1
Solution Test
10% v/v in diethyl ether : Clear
Solution Test
10% v/v in methanol : Clear
Appearance (Color)
Clear colorless
Assay (GC)
≥97.5%
Refractive Index
1.5110-1.5150 @ 20?C

Description

Benzyl methacrylate is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzyl methacrylate is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. It is sensitive to light and heat. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text