3-(Trifluoromethyl)phenylacetic acid, 96%
3-(Trifluoromethyl)phenylacetic acid, 96%
3-(Trifluoromethyl)phenylacetic acid, 96%
Thermo Scientific Chemicals

3-(Trifluoromethyl)phenylacetic acid, 96%

CAS: 351-35-9 | C9H7F3O2 | 204.148 g/mol
製品番号(カタログ番号) L10960.18
または、製品番号L10960-18
価格(JPY)
-
数量:
50 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS351-35-9
IUPAC Name2-[3-(trifluoromethyl)phenyl]acetic acid
Molecular FormulaC9H7F3O2
InChI KeyBLXXCCIBGGBDHI-UHFFFAOYSA-N
SMILESOC(=O)CC1=CC=CC(=C1)C(F)(F)F
さらに表示
Assay (Silylated GC)≥95.0%
FormCrystals or powder or crystalline powder or lumps or chunks or fused solid
Identification (FTIR)Conforms
Melting Point (clear melt)71.0-81.0?C
Appearance (Color)White to cream to pale brown
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only