Thermo Scientific Chemicals

4-Iodo-3-nitroanisole, 98+%, Thermo Scientific Chemicals

Catalog number: L11714.14
25 g, Each
Thermo Scientific Chemicals

4-Iodo-3-nitroanisole, 98+%, Thermo Scientific Chemicals

Catalog number: L11714.14
25 g, Each
Quantity
Catalog number: L11714.14
also known as L11714-14
Price (USD)
Price: 252.00
Online price: 217.65
Your price:
Quantity
-

Chemical Identifiers

CAS
58755-70-7
IUPAC Name
1-iodo-4-methoxy-2-nitrobenzene
Molecular Formula
C7H6INO3
InChI Key
JWZODIRSJJQOKY-UHFFFAOYSA-N
SMILES
COC1=CC=C(I)C(=C1)[N+]([O-])=O
Melting Point (clear melt)
58.0-65.0?C
Appearance (Color)
Yellow to orange to dark red to red to brown
Assay (GC)
≥98.0%
Form
Crystals or powder or crystalline powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text