1-Decylphosphonic acid, 98%
1-Decylphosphonic acid, 98%
1-Decylphosphonic acid, 98%
Thermo Scientific Chemicals

1-Decylphosphonic acid, 98%

CAS: 6874-60-8 | C10H23O3P | 222.265 g/mol
製品番号(カタログ番号) L12019.03
または、製品番号L12019-03
価格(JPY)
-
数量:
1 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS6874-60-8
IUPAC Namedecylphosphonic acid
Molecular FormulaC10H23O3P
InChI KeyDZQISOJKASMITI-UHFFFAOYSA-N
SMILESCCCCCCCCCCP(O)(O)=O
さらに表示
Assay (Silylated GC)≥97.5%
FormCrystals or powder or crystalline powder or waxy solid
Melting Point (clear melt)98.0-104.0?C
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
Appearance (Color)White
1-Decylphosphonic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff. It is used in stabilizing dye-sensitized solar cell.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Decylphosphonic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff. It is used in stabilizing dye-sensitized solar cell.

Notes
Store in a cool, dry conditions in well sealed container. Incompatible with oxidizing agents.
RUO – Research Use Only
P. Wang; S.M. Zakeeruddin; R. Humphry-Baker; J.E. Moser; M. Grätzel. Molecular-Scale Interface Engineering of TiO2 Nanocrystals: Improve the Efficiency and Stability of Dye-Sensitized Solar Cells. Communication.2003, 15, 2101-2104.