1-Bromo-2-butanone, tech. 90%, stab. with calcium carbonate, Thermo Scientific Chemicals
1-Bromo-2-butanone, tech. 90%, stab. with calcium carbonate, Thermo Scientific Chemicals
1-Bromo-2-butanone, tech. 90%, stab. with calcium carbonate, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1-Bromo-2-butanone, tech. 90%, stab. with calcium carbonate, Thermo Scientific Chemicals

 
製品番号(カタログ番号) L12081.06
価格(JPY)
-
数量:
5 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS816-40-0
IUPAC Name1-bromobutan-2-one
Molecular FormulaC4H7BrO
InChI KeyCCXQVBSQUQCEEO-UHFFFAOYSA-N
SMILESCCC(=O)CBr
さらに表示
CommentProduct contains white sediment or suspension, filter before use.
Appearance (Color)Colorless to yellow
StabilizerApprox. 0.5 % CaCO3
FormLiquid
Assay (GC)≥88.0%
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
 
RUO – Research Use Only