5-Chloro-1-pentanol, 95%
5-Chloro-1-pentanol, 95%
5-Chloro-1-pentanol, 95%
Thermo Scientific Chemicals

5-Chloro-1-pentanol, 95%

CAS: 5259-98-3 | C5H11ClO | 122.59 g/mol
製品番号(カタログ番号) L12088.18
または、製品番号L12088-18
価格(JPY)
-
数量:
50 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS5259-98-3
Appearance (Color)Clear colorless to pale yellow
Refractive Index1.4510-1.4570 @ 20°C
FormLiquid or viscous liquid
Assay (GC)≥92.0%
5-Chloro-1-pentanol is used to produce 6-hydroxy-hexanenitrile at temperature of 90 - 100°C. It is also is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Chloro-1-pentanol is used to produce 6-hydroxy-hexanenitrile at temperature of 90 - 100°C. It is also is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

Solubility
Slightly soluble in water.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only