Thermo Scientific Chemicals

3-Diethylamino-1-propyne, 98+%, Thermo Scientific Chemicals

Catalog number: L12237.14
25 g, Each
Thermo Scientific Chemicals

3-Diethylamino-1-propyne, 98+%, Thermo Scientific Chemicals

Catalog number: L12237.14
25 g, Each
Quantity
Catalog number: L12237.14
also known as L12237-14
Price (USD)
Price: 211.00
Online price: 181.65
Your price:
Quantity
-

Chemical Identifiers

CAS
4079-68-9
IUPAC Name
diethyl(prop-2-yn-1-yl)amine
Molecular Formula
C7H13N
InChI Key
JZJXKEWVUBVOEH-UHFFFAOYSA-N
SMILES
CCN(CC)CC#C
Appearance (Color)
Clear colorless to yellow
Assay (GC)
≥98.0%
Form
Liquid
Refractive Index
1.4300-1.4340 @ 20?C

Description

3-Diethylamino-1-propyne is used as functional leveling agent, brightener.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Diethylamino-1-propyne is used as functional leveling agent, brightener.

Solubility
Soluble in acid. Insoluble in water.

Notes
Incompatible with heat, light, carbondioxide, heavy metal powders. Air & Light Sensitive. Store in the dark. Store under dry inert gas.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text