2-Hydroxythiophenol, 97%, Thermo Scientific Chemicals
2-Hydroxythiophenol, 97%, Thermo Scientific Chemicals
2-Hydroxythiophenol, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Hydroxythiophenol, 97%, Thermo Scientific Chemicals

製品番号(カタログ番号)数量
L12519.06
または、製品番号L12519-06
5 g
製品番号(カタログ番号) L12519.06
または、製品番号L12519-06
価格(JPY)
-
数量:
5 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS1121-24-0
IUPAC Name2-sulfanylphenol
Molecular FormulaC6H6OS
InChI KeyVMKYTRPNOVFCGZ-UHFFFAOYSA-N
SMILESOC1=CC=CC=C1S
さらに表示
Appearance (Color)Clear, colorless to yellow
Assay from Suppliers CofA≥96.0%
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only