Ethyl 2-isothiocyanatopropionate, 97%
Ethyl 2-isothiocyanatopropionate, 97%
Ethyl 2-isothiocyanatopropionate, 97%
Thermo Scientific Chemicals

Ethyl 2-isothiocyanatopropionate, 97%

CAS: 39574-16-8 | C6H9NO2S | 159.20 g/mol
製品番号(カタログ番号) L12532.03
または、製品番号L12532-03
価格(JPY)
-
数量:
1 g
一括またはカスタム形式をリクエストする
仕様
バイルシュタイン1723243
CAS39574-16-8
化学物質名または材質Ethyl 2-isothiocyanatopropionate
DOT情報Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.
式量159.2
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only