1,4-Butanediol diglycidyl ether, 96%
1,4-Butanediol diglycidyl ether, 96%
1,4-Butanediol diglycidyl ether, 96%
Thermo Scientific Chemicals

1,4-Butanediol diglycidyl ether, 96%

CAS: 2425-79-8 | C10H18O4 | 202.25 g/mol
製品番号(カタログ番号) L12632.09
または、製品番号L12632-09
価格(JPY)
-
数量:
10 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS2425-79-8
IUPAC Name2-({4-[(oxiran-2-yl)methoxy]butoxy}methyl)oxirane
Molecular FormulaC10H18O4
InChI KeySHKUUQIDMUMQQK-UHFFFAOYNA-N
SMILESC(CCOCC1CO1)COCC1CO1
さらに表示
Appearance (Color)Clear colorless to yellow
Refractive Index1.4510-1.4585 @ 20°C (non-U.S. specification)
FormLiquid
Assay (GC)≥95.0%
CommentSpecification differs for U.S. and non-U.S. material where indicated
さらに表示
1,4-Butanediol diglycidyl ether is used for preparing amylose, xylan and hydroxyethyl-cellulose, to cross link polyethylenimine which also serves as a hole-blocking layer in organic solar cells, to cross link hyaluronic acid into hydrogels and for the activation of soluble dextran polymers. It is the most commonly used homobifunctional epoxide compound

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,4-Butanediol diglycidyl ether is used for preparing amylose, xylan and hydroxyethyl-cellulose, to cross link polyethylenimine which also serves as a hole-blocking layer in organic solar cells, to cross link hyaluronic acid into hydrogels and for the activation of soluble dextran polymers. It is the most commonly used homobifunctional epoxide compound

Solubility
Soluble in water (55,6 g/L at 20°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents. Store at room temperature.
RUO – Research Use Only

General References:

  1. Leonid N Ten.; Wan-Taek Im.; Myung-Kyum Kim.; Myung Suk Kang.; Sung-Taik Lee. Development of a plate technique for screening of polysaccharide-degrading microorganisms by using a mixture of insoluble chromogenic substrates. Journal of Microbiological Methods. 2004, 56 (3), 375-382.
  2. Lennart Kenne.; Suresh Gohil.; Eva M Nilsson.; Anders Karlsson.; David Ericsson.; Anne Helander Kenne.; Lars I Nord. Modification and cross-linking parameters in hyaluronic acid hydrogels--definitions and analytical methods. Carbohydrate polymers. 2013, 91 (1), 410-418.