Thermo Scientific Chemicals

4-Ethylphenyl isocyanate, 98%, Thermo Scientific Chemicals

Catalog number: L12718.14
25 g, Each
Thermo Scientific Chemicals

4-Ethylphenyl isocyanate, 98%, Thermo Scientific Chemicals

Catalog number: L12718.14
25 g, Each
Quantity
Catalog number: L12718.14
also known as L12718-14
Price (USD)
Price: 184.00
Online price: 158.65
Your price:
Quantity
-

Chemical Identifiers

CAS
23138-50-3
IUPAC Name
1-ethyl-4-isocyanatobenzene
Molecular Formula
C9H9NO
InChI Key
FWPYUSLQCQDLJR-UHFFFAOYSA-N
SMILES
CCC1=CC=C(C=C1)N=C=O
Appearance (Color)
Clear or slightly turbid colorless to pale yellow
Assay (GC)
≥97.5%
Form
Liquid
Comment
May form slight sediment on storage
Refractive Index
1.5250-1.5290 @ 20?C

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text