Diethyl 1-decylphosphonate, 97%
Diethyl 1-decylphosphonate, 97%
Diethyl 1-decylphosphonate, 97%
Diethyl 1-decylphosphonate, 97%
Thermo Scientific Chemicals

Diethyl 1-decylphosphonate, 97%

CAS: 16165-68-7 | C14H31O3P | 278.373 g/mol
製品番号(カタログ番号) L13038.14
または、製品番号L13038-14
価格(JPY)
-
見積もりを依頼する
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS16165-68-7
IUPAC Namediethyl decylphosphonate
Molecular FormulaC14H31O3P
InChI KeyHYQXIVAZQBYBAF-UHFFFAOYSA-N
SMILESCCCCCCCCCCP(=O)(OCC)OCC
さらに表示
Appearance (Color)Clear colorless
Assay (GC)≥96.0%
Refractive Index1.4350-1.4400 @ 20?C
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only