Sodium ethoxide, 21% w/w in ethanol
Sodium ethoxide, 21% w/w in ethanol
Sodium ethoxide, 21% w/w in ethanol
Sodium ethoxide, 21% w/w in ethanol
Thermo Scientific Chemicals

Sodium ethoxide, 21% w/w in ethanol

CAS: 141-52-6 | C2H5NaO | 68.051 g/mol
数量:
100 mL
500 mL
2500 mL
製品番号(カタログ番号) L13083.AE
または、製品番号L13083-AE
価格(JPY)
-
見積もりを依頼する
数量:
100 mL
一括またはカスタム形式をリクエストする
化学物質識別子
CAS141-52-6
IUPAC Namesodium ethanolate
Molecular FormulaC2H5NaO
InChI KeyQDRKDTQENPPHOJ-UHFFFAOYSA-N
SMILES[Na+].CC[O-]
さらに表示
Appearance (Color)Clear pale yellow to orange to brown
FormLiquid
Assay (Non-aqueous acid-base Titration)≥20.0 to ≤22.0% (Sodium ethoxide)
Impurity content≤0.50% [NaOH + Na2CO3 (as NaOH)]

Sodium ethoxide, 21% w/w in ethanol is used as a strong base in organic synthesis. It finds application in various chemical reactions such as condensation, esterification, alkoxylation and etherifcation. It is actively involved in Claisen condensation, Stobbe reaction and Wolf-kishner reduction. It is an important starting material for the synthesis of ethyl ester and diethyl ester of malonic acid. In Williamson ether synthesis, it reacts with ethyl bromide to form diethyl ether.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium ethoxide, 21% w/w in ethanol is used as a strong base in organic synthesis. It finds application in various chemical reactions such as condensation, esterification, alkoxylation and etherifcation. It is actively involved in Claisen condensation, Stobbe reaction and Wolf-kishner reduction. It is an important starting material for the synthesis of ethyl ester and diethyl ester of malonic acid. In Williamson ether synthesis, it reacts with ethyl bromide to form diethyl ether.

Solubility
Miscible with alcohol.

Notes
Moisture sensitive. It hydrolyzes rapidly into sodium hydroxide in the presence of moist air. Incompatible with water, strong bases, acids, oxidizing agents, alkali metals, strong oxidizing agents, ammonia, acid chlorides, acid anhydrides, reducing agents, peroxides, acids and chlorinated solvents.
For Manufacturing and Laboratory Use. Not for beverage use

General References:

  1. Abduh, M. Y.; van Ulden, W.; Kalpoe, V.; van de Bovenkamp, H. H.; Manurung, R.; Heeres, H. J. Biodiesel synthesis from Jatropha curcas L. oil and ethanol in a continuous centrifugal contactor separator. Eur. J. Lipid Sci. Technol. 2013, 115 (1), 123-131.
  2. Parsons, R. V.; Cenkowski, S.; Sorensen, J. L.; Beta, T.; Arntfield, S. D. Hemicellulose polysaccharide recovery from flax shive using alkaline solutions with sodium ethoxide pretreatment. Ind. Crops Prod. 2013, 44, 165-170.