L-Leucine ethyl ester hydrochloride, 97%
L-Leucine ethyl ester hydrochloride, 97%
L-Leucine ethyl ester hydrochloride, 97%
Thermo Scientific Chemicals

L-Leucine ethyl ester hydrochloride, 97%

CAS: 2743-40-0 | C8H17NO2 | 159.23 g/mol
製品番号(カタログ番号) L13147.14
または、製品番号L13147-14
価格(JPY)
-
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS2743-40-0
IUPAC Nameethyl (2S)-2-amino-4-methylpentanoate
Molecular FormulaC8H17NO2
InChI KeyQIGLJVBIRIXQRN-ZETCQYMHSA-N
SMILESCCOC(=O)[C@@H](N)CC(C)C
さらに表示
Appearance (Color)White
FormCrystals or powder or crystalline powder or fused solid or needles
Assay (Titration ex Chloride)≥96.0 to ≤104.0%
Melting Point (clear melt)134-138°C
Optical Rotation+16.5° to +19.5° (c=5 in ethanol)
L-Leucine ethyl ester hydrochloride is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Leucine ethyl ester hydrochloride is used as an organic chemical synthesis intermediate.

Solubility
Soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. K.S.Chio; Aloys L.Tappel. Synthesis and characterization of the fluorescent products derived from malonaldehyde and amino acids. Biochemistry. 1969, 8,(7), 2821-2827.
  2. Robert L.Letsinger; Milton J.Kornet. Popcorn polymer as a support in multistep syntheses. J. Am. Chem. Soc. 1963, 85,(19), 3045-3046.