Zinc powder, -100 mesh, 97+%, Thermo Scientific Chemicals
Zinc powder, -100 mesh, 97+%, Thermo Scientific Chemicals
Zinc powder, -100 mesh, 97+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Zinc powder, -100 mesh, 97+%, Thermo Scientific Chemicals

数量:
100 g
500 g
2.5 kg
製品番号(カタログ番号) L13310.22
価格(JPY)
-
数量:
100 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS7440-66-6
IUPAC Namezinc
Molecular FormulaZn
InChI KeyHCHKCACWOHOZIP-UHFFFAOYSA-N
SMILES[Zn]
さらに表示
Appearance (Color)Grey
FormPowder
Assay (unspecified)≥97.0 to ≤103.0% (U.S. specification)
CommentSpecification differs for U.S. and non-U.S. material where indicated
Particle Size-100 mesh (<150 microns)
Zinc powder is used in thermal spraying for corrosion protection and in the chemical industry. It provides chemical potential energy in alkaline batteries to pump electrical charges between the negative and positive terminals. It is used as a fuel for model rockets with a mixture of 2.04 parts zinc to one part sulfur. It is used as protective pigment in industrial coatings, as a reducing agent and catalyst in a variety of chemical reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Zinc powder is used in thermal spraying for corrosion protection and in the chemical industry. It provides chemical potential energy in alkaline batteries to pump electrical charges between the negative and positive terminals. It is used as a fuel for model rockets with a mixture of 2.04 parts zinc to one part sulfur. It is used as protective pigment in industrial coatings, as a reducing agent and catalyst in a variety of chemical reactions.

Solubility
Reacts with water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store under dry inert gas. It is sensitive to air and moisture. Incompatible with strong acids, strong bases and oxidizing agents.
RUO – Research Use Only

General References:

  1. Anna M. Polcaro.; Simona Palmas.; Stella Dernini. Kinetics of cobalt cementation on zinc powder. Ind. Eng. Chem. 1995, 34 (9), 3090-3095.
  2. Mihai Rusua.; Nicoleta Sofiana.; Daniela Rusu. Mechanical and thermal properties of zinc powder filled high density polyethylene composites. Polymer Testing. 2001, 20 (4), 409-417.