1-Dodecyl isocyanate, 98%
1-Dodecyl isocyanate, 98%
1-Dodecyl isocyanate, 98%
Thermo Scientific Chemicals

1-Dodecyl isocyanate, 98%

CAS: 4202-38-4 | C13H25NO | 211.349 g/mol
製品番号(カタログ番号) L13380.03
または、製品番号L13380-03
価格(JPY)
-
数量:
1 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS4202-38-4
IUPAC Name1-isocyanatododecane
Molecular FormulaC13H25NO
InChI KeyYIDSTEJLDQMWBR-UHFFFAOYSA-N
SMILESCCCCCCCCCCCCN=C=O
さらに表示
Assay (GC)> 97.5%
Appearance (Color)Colourless
FormClear to slightly turbid liquid
CommentMay form slight sediment on storage, filter before use.
Refractive Index1.4385 - 1.4435 @20?C
1-Dodecyl isocyanate, is incorporated in AZT analogue preparation through it use ine the synthesis of carbodiimides from phosphinimines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Dodecyl isocyanate, is incorporated in AZT analogue preparation through it use ine the synthesis of carbodiimides from phosphinimines.

Solubility
It hydrolyzes in water.

Notes
Moisture Sensitive. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
RUO – Research Use Only

General References:

  1. SL Huang.; MS Chao.; RC Ruaan.;JY Lai. Microphase separated structure and protein adsorption of polyurethanes with butadiene soft segment. European Polymer Journal. 2000, 36,(2), 285-294.
  2. AR Doumaux Jr. Marked differences between the sodium-ammonia and calcium-ammonia reduction of nitriles. The Journal of Organic Chemistry. 1972, 37,(3), 508-510.