5(6)-Carboxyfluorescein, mixture of isomers, 97%
5(6)-Carboxyfluorescein, mixture of isomers, 97%
5(6)-Carboxyfluorescein, mixture of isomers, 97%
Thermo Scientific Chemicals

5(6)-Carboxyfluorescein, mixture of isomers, 97%

CAS: 72088-94-9 | C42H24O14 | 752.64 g/mol
製品番号(カタログ番号) L13439.MD
または、製品番号L13439-MD
価格(JPY)
-
数量:
250 mg
一括またはカスタム形式をリクエストする
化学物質識別子
CAS72088-94-9
Appearance (Color)Orange to brown or orange-red or orange-brown
FormPowder or crystalline powder and/or chunks
Assay (HPLC)≥96.0% (sum of isomers)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only