AmberChrom™, 50WX8 200-400 (H)
AmberChrom™, 50WX8 200-400 (H)
AmberChrom™, 50WX8 200-400 (H)
AmberChrom™, 50WX8 200-400 (H)
Thermo Scientific Chemicals

AmberChrom™, 50WX8 200-400 (H)

CAS: 11119-67-8
数量:
50 g
250 g
1000 g
製品番号(カタログ番号) L13922.18
または、製品番号L13922-18
価格(JPY)
-
見積もりを依頼する
数量:
50 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS11119-67-8
IUPAC NameAmberChrom™ 50WX8 Ion Exchange Resin
MDL NumberMFCD00132726
FormIonic form: H⁺
Moisture holding capacity50-58% (H)
Particle Size-325 mesh (<44 micron) : ≤15%
CommentStrongly acidic cation exchange resin.
MatrixPolystyrene crosslinked with 8% divinylbenzene with sulphonic acid functional groups
さらに表示

AmberChrome™ 50WX8 Ion Exchange Resin) is used in separation medium for TLC and LPLC. It is also used as stripper resin and can be regenerated by 0.25N Sulfuricacid when exhausted, is used as catalyst and in deionizing supernatant fraction samples for paper chromatography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
AmberChrome™ 50WX8 Ion Exchange Resin is used in separation medium for TLC and LPLC. It is also used as stripper resin and can be regenerated by 0.25N Sulfuricacid when exhausted, is used as catalyst and in deionizing supernatant fraction samples for paper chromatography.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Small, Hamish.; Timothy S. Stevens.; William C. Bauman. Novel ion exchange chromatographic method using conductimetric detection. Anal. Chem. 1975, 47 1801-1809.
  2. Claverie-Martin.; Felix, Maria R. Diaz-Torres.; Michael J. Geoghegan. Chemical composition and ultrastructure of wild-type and white mutantAspergillus nidulans conidial walls. Curr. Microbiol. 1988, 16 281-287.
  3. Strongly acidic cation-exchange resin, 8% cross-linked, H+ form, 200-400 mesh (dry).