Thermo Scientific Chemicals

N-Acetyl-L-leucine, 99%

Catalog number: L13926.06
5 g, Each
Thermo Scientific Chemicals

N-Acetyl-L-leucine, 99%

Catalog number: L13926.06
5 g, Each
Quantity
Catalog number: L13926.06
also known as L13926-06
Price (USD)

Chemical Identifiers

CAS
1188-21-2
IUPAC Name
(2S)-2-acetamido-4-methylpentanoic acid
Molecular Formula
C8H15NO3
InChI Key
WXNXCEHXYPACJF-JLDDOWRYNA-N
SMILES
CC(C)C[C@H](NC(C)=O)C(O)=O
Appearance (Color)
White
Form
Crystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)
≥97.5 to ≤102.5% (non-U.S. specification)
Assay from Suppliers CofA
≥97.5% (U.S. specification)
Comment
Specification differs for U.S. and non-U.S. material where indicated

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text