N-Phenyldiethanolamine, 97%
N-Phenyldiethanolamine, 97%
N-Phenyldiethanolamine, 97%
Thermo Scientific Chemicals

N-Phenyldiethanolamine, 97%

CAS: 120-07-0 | C10H15NO2 | 181.24 g/mol
数量:
100 g
500 g
2500 g
製品番号(カタログ番号) L13931.36
または、製品番号L13931-36
価格(JPY)
-
数量:
500 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS120-07-0
IUPAC Name2-[(2-hydroxyethyl)(phenyl)amino]ethan-1-ol
Molecular FormulaC10H15NO2
InChI KeyOJPDDQSCZGTACX-UHFFFAOYSA-N
SMILESOCCN(CCO)C1=CC=CC=C1
さらに表示
Melting Point (clear melt)54.0-62.0°C
Appearance (Color)White to cream to dark yellow to pale brown or blue to purple
FormCrystals or powder or crystalline powder or lumps or chunks or fused solid
Assay (GC)≥96.0%
Identification (FTIR)Conforms
N-Phenyldiethanolamine is used as phenylephrine intermediates, used as dye intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Phenyldiethanolamine is used as phenylephrine intermediates, used as dye intermediates.

Solubility
Soluble in water 3g/L @ 20˚C.

Notes
Air sensitive & hygroscopic. Store under inert gas. Store away from oxidizing agents, air, water/ moisture.
RUO – Research Use Only

General References:

  1. Paul B. Hodgson; Fabrice H. Salingue. The preparation of a stable 2-pyridylboronate and its reactivity in the Suzuki-Miyaura cross-coupling reaction. Tetrahedron Letters. 2004, 45, (4), 685-687.
  2. Yoshiaki Shirae; Takashi Mino; Tae Hasegawa; Masami Sakamoto;Tsutomu Fujita. Transesterification of various alcohols with vinyl acetate under mild conditions catalyzed by diethylzinc using N-substituted diethanolamine as a ligand. Tetrahedron Letters. 1987, 46, (35), 5877-5879.