Thermo Scientific Chemicals

Amberchrom™ 50Wx2 100-200 (H), Thermo Scientific Chemicals

Catalog number: L13944.30
250 g, Each
Thermo Scientific Chemicals

Amberchrom™ 50Wx2 100-200 (H), Thermo Scientific Chemicals

Catalog number: L13944.30
250 g, Each
Quantity
Catalog number: L13944.30
also known as L13944-30
Price (USD)
Price: 174.00
Online price: 147.65
Your price:
Quantity
-

Chemical Identifiers

CAS
12612-37-2
MDL Number
MFCD00132722
Form
Beads
Exchange capacity
≥0.6 eq/L (as H+ form)
Comment
Strongly acidic cation exchange resin, ionic form : H+
Moisture holding capacity
74.0-82.0% (as H+ form)
Particle Size
>500 micron (35mesh): ≤15.0%

Description

Dowex™ 50WX2 100-200 (H) is used to remove calcium from aqueous samples. It plays a good role in drug delivery systems.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Amberchrom 50WX2 100-200 (H) is used to remove calcium from aqueous samples. It plays a good role in drug delivery systems.

Solubility
Insoluble in water.

Notes
Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text