tert-Butyl methyl ether, 99%
tert-Butyl methyl ether, 99%
tert-Butyl methyl ether, 99%
Thermo Scientific Chemicals

tert-Butyl methyl ether, 99%

CAS: 1634-04-4 | C5H12O | 88.15 g/mol
数量:
500 mL
1000 mL
2500 mL
製品番号(カタログ番号) L14030.AU
または、製品番号L14030-AU
価格(JPY)
-
数量:
1000 mL
一括またはカスタム形式をリクエストする
化学物質識別子
CAS1634-04-4
IUPAC Name2-methoxy-2-methylpropane
Molecular FormulaC5H12O
InChI KeyBZLVMXJERCGZMT-UHFFFAOYSA-N
SMILESCOC(C)(C)C
さらに表示
Assay (GC)≥98.5% (non-U.S. specification)
Assay from Suppliers CofA≥98.5% (GC: U.S. specification)
FormLiquid
CommentSpecification differs for U.S. and non-U.S. material where indicated
Identification (FTIR)Conforms (non-U.S. specification)
さらに表示
Used as a chromatography solvent. Acts as a fuel component in fuel for gasoline engines. They raise the oxygen content of gasoline(oxygenates).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as a chromatography solvent. Acts as a fuel component in fuel for gasoline engines. They raise the oxygen content of gasoline(oxygenates).

Solubility
Miscible with ethanol and ethyl ether. Immiscible with water.

Notes
Store in a cool, dry, well-ventilated area. It is hygroscopic in nature.
RUO – Research Use Only

General References:

  1. Fischer, A.; Oehm, C.; Selle, M.; Werner, P.; Oehm; Selle; Werner. Biotic and abiotic transformations of methyl tertiary butyl ether (MTBE). Environ. Sci. Pollut. Res. Int. 2005, 12 (6), 381-389.
  2. Matyash, V.; Liebisch, G.; Kurzchalia, T. V.; Shevchenko, A.; Schwudke, D. Lipid extraction by methyl-tert-butyl ether for high-throughput lipidomics. J. Lipid Res. 2008, 49 (5), 1137-1146.