delta-Nonanolactone, 98%, Thermo Scientific Chemicals
delta-Nonanolactone, 98%, Thermo Scientific Chemicals
delta-Nonanolactone, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

delta-Nonanolactone, 98%, Thermo Scientific Chemicals

製品番号(カタログ番号) L14122.22
または、製品番号L14122-22
価格(JPY)
-
数量:
100 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS3301-94-8
IUPAC Name6-butyloxan-2-one
Molecular FormulaC9H16O2
InChI KeyPXRBWNLUQYZAAX-UHFFFAOYNA-N
SMILESCCCCC1CCCC(=O)O1
さらに表示
Assay (GC)≥97.5%
Refractive Index1.4555-1.4595 @ 20?C
Appearance (Color)Clear colorless
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only