Pyridinium dichromate, 98%
Pyridinium dichromate, 98%
Pyridinium dichromate, 98%
Thermo Scientific Chemicals

Pyridinium dichromate, 98%

Alfa Aesar Pyridinium dichromate, 98%, Quantity: 100g, Melting Point: ∼145 deg.C to 147 deg.C, Molecular Weight: 376.205, Percent Purity: 98%, CAS: 20039-37-6, DOT Information: Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: OXIDIZING SOLID, TOXIC, N.O.S. | CAS: 20039-37-6 | C10H12Cr2N2O7 | 376.205 g/mol
製品番号(カタログ番号)数量
L15132.14
または、製品番号L15132-14
25 g
製品番号(カタログ番号) L15132.14
または、製品番号L15132-14
価格(JPY)
-
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS20039-37-6
IUPAC Name[(hydroxydioxochromio)oxy]chromiumoylol; bis(pyridine)
Molecular FormulaC10H12Cr2N2O7
InChI KeyRCBVKBFIWMOMHF-UHFFFAOYSA-L
SMILESC1=CC=NC=C1.C1=CC=NC=C1.O[Cr](=O)(=O)O[Cr](O)(=O)=O
さらに表示
Appearance (Color)Orange to brown
Assay (Iodometric Titration)≥97.5 to ≤102.5%
FormCrystals or powder or crystalline powder
Identification (FTIR)Conforms
Pyridinium dichromate acts as a strong oxidizing agent used in the conversion of primary alcohols and secondary alcohols to aldehydes and ketones respectively. It plays an important role in the oxidation of unsaturated tertiary alcohols, silyl ethers, the carbon-boron bond, and oximes. Further, it is used in the conversion of acetals to esters and didehydroketones to enones in the presence of tert-butyl hydroperoxide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pyridinium dichromate acts as a strong oxidizing agent used in the conversion of primary alcohols and secondary alcohols to aldehydes and ketones respectively. It plays an important role in the oxidation of unsaturated tertiary alcohols, silyl ethers, the carbon-boron bond, and oximes. Further, it is used in the conversion of acetals to esters and didehydroketones to enones in the presence of tert-butyl hydroperoxide.

Solubility
Soluble in water, dimethyl formamide, dimethyl sulfoxide and acetonitrile. Slightly soluble in dichloromethane, chloroform and acetone. Insoluble in hexane, ether and ethyl acetate.

Notes
Hygroscopic. Incompatible withorganic materials and powdered metals.
RUO – Research Use Only

General References:

  1. Selective oxidant which can be used in dichloromethane to convert primary alcohols to aldehydes: Tetrahedron Lett., 399 (1979). Can also be used catalytically in combination with Sodium percarbonate, A16045, as co-oxidant for conversion of allylic and benzylic alcohols to aldehydes under phase-transfer conditions: Tetrahedron Lett., 35, 1989 (1994). Under the same conditions, ɑ-functionalized benzylic alcohols undergo oxidative cleavage: Synth. Commun., 25, 2051 (1995).
  2. Matsunaga, K.; Hirajima, H.; Kishida, A.; Takatori, K.; Nagaoka, H. Novel PDC catalyzed oxidative rearrangement of tertiary allylic alcohols to beta-substituted enones. Tetrahedron Lett. 2015, 56 (43), 5941-5944.
  3. Ghosh, A.; Saha, R.; Mukherjee, K.; Ghosh, S. K.; Sar, P.; Malik, S.; Saha, B. Choice of suitable micellar catalyst for 2,2'-bipyridine-promoted chromic acid oxidation of glycerol to glyceraldehyde in aqueous media at room temperature. Res. Chem. Intermed. 2015, 41 (5), 3057-3078.