4,5-Dibromo-2-furoic acid, 97%
4,5-Dibromo-2-furoic acid, 97%
4,5-Dibromo-2-furoic acid, 97%
Thermo Scientific Chemicals

4,5-Dibromo-2-furoic acid, 97%

CAS: 2434-03-9 | C5H2Br2O3 | 269.88 g/mol
製品番号(カタログ番号) L15517.06
または、製品番号L15517-06
価格(JPY)
-
数量:
5 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS2434-03-9
IUPAC Name4,5-dibromofuran-2-carboxylic acid
Molecular FormulaC5H2Br2O3
InChI KeyBHUVICYZDBUMIU-UHFFFAOYSA-N
SMILESOC(=O)C1=CC(Br)=C(Br)O1
さらに表示
FormPowder or crystalline powder
Assay (Aqueous acid-base Titration)≥96.0 to ≤104.0%
Melting Point (clear melt)167-173?C
Appearance (Color)White to cream to pale brown
Assay (HPLC)≥96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only