Cycloheptylacetic acid, 99%
Cycloheptylacetic acid, 99%
Cycloheptylacetic acid, 99%
Thermo Scientific Chemicals

Cycloheptylacetic acid, 99%

CAS: 4401-20-1 | C9H16O2 | 156.225 g/mol
製品番号(カタログ番号) L15553.03
または、製品番号L15553-03
価格(JPY)
-
数量:
1 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS4401-20-1
IUPAC Name2-cycloheptylacetic acid
Molecular FormulaC9H16O2
InChI KeyDQNWKASPZFJVMJ-UHFFFAOYSA-N
SMILESOC(=O)CC1CCCCCC1
さらに表示
Assay (Aqueous acid-base Titration)≥98.5 to ≤101.5%
Assay (GC)≥98.5%
Refractive Index1.4705-1.4735 @ 20?C
Appearance (Color)Clear colorless
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only