1,4-Diphenyl-1-butanone, 98%
1,4-Diphenyl-1-butanone, 98%
1,4-Diphenyl-1-butanone, 98%
Thermo Scientific Chemicals

1,4-Diphenyl-1-butanone, 98%

CAS: 5407-91-0 | C16H16O | 224.303 g/mol
製品番号(カタログ番号) L15597.06
または、製品番号L15597-06
価格(JPY)
-
数量:
5 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS5407-91-0
IUPAC Name1,4-diphenylbutan-1-one
Molecular FormulaC16H16O
InChI KeyGBUMEGLMTNAXOM-UHFFFAOYSA-N
SMILESO=C(CCCC1=CC=CC=C1)C1=CC=CC=C1
さらに表示
FormPowder or granules
Assay (GC)≥97.5%
Melting Point (clear melt)51.5-57.5?C
Appearance (Color)White to pale yellow or pale pink
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only