Thermo Scientific Chemicals

4-Chloro-3-nitro-2-pyridone, 98%, Thermo Scientific Chemicals

Catalog number: L15686.14
25 g, Each
Thermo Scientific Chemicals

4-Chloro-3-nitro-2-pyridone, 98%, Thermo Scientific Chemicals

Catalog number: L15686.14
25 g, Each
Quantity
Catalog number: L15686.14
also known as L15686-14
Price (USD)
Quantity
-

Chemical Identifiers

CAS
165547-79-5
IUPAC Name
4-chloro-3-nitro-1,2-dihydropyridin-2-one
Molecular Formula
C5H3ClN2O3
InChI Key
UKIZCTHOMJXNIX-UHFFFAOYSA-N
SMILES
[O-][N+](=O)C1=C(Cl)C=CNC1=O
Melting Point (clear melt)
219.5-225.5?C
Appearance (Color)
Yellow
Form
Powder
Assay (HPLC)
≥97.5%

Description

4-Chloro-3-nitro-2-pyridone finds an important application as raw material in organic synthesis and pharmaceuticals. It is also used as an intermediate in agrochemicals and dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Chloro-3-nitro-2-pyridone finds an important application as raw material in organic synthesis and pharmaceuticals. It is also used as an intermediate in agrochemicals and dyestuffs.

Solubility
Insoluble in water.

Notes
Keep the container tightly sealed. Store in a cool, dry condition. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text