Tri(4-morpholinyl)phosphine oxide, 99%
Tri(4-morpholinyl)phosphine oxide, 99%
Tri(4-morpholinyl)phosphine oxide, 99%
Tri(4-morpholinyl)phosphine oxide, 99%
Thermo Scientific Chemicals

Tri(4-morpholinyl)phosphine oxide, 99%

CAS: 4441-12-7 | C24H42 | 330.60 g/mol
製品番号(カタログ番号) L16315.14
または、製品番号L16315-14
価格(JPY)
-
見積もりを依頼する
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS4441-12-7
IUPAC Name4-[bis(morpholin-4-yl)phosphoroso]morpholine
Molecular FormulaC12H24N3O4P
InChI KeyWXMQHPKQCPCDQO-UHFFFAOYSA-N
SMILESO=P(N1CCOCC1)(N1CCOCC1)N1CCOCC1
さらに表示
Assay (GC)≥98.5%
Appearance (Color)White to cream to pale yellow
FormCrystals or powder or crystalline powder or lumps or fused solid or needles
Identification (FTIR)Conforms
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only