Heptafluoro-2-iodopropane, 97%, stab. with copper powder
Heptafluoro-2-iodopropane, 97%, stab. with copper powder
Heptafluoro-2-iodopropane, 97%, stab. with copper powder
Thermo Scientific Chemicals

Heptafluoro-2-iodopropane, 97%, stab. with copper powder

CAS: 677-69-0 | C3F7I | 295.926 g/mol
数量:
5 g
25 g
100 g
製品番号(カタログ番号) L16523.06
または、製品番号L16523-06
価格(JPY)
-
数量:
5 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS677-69-0
IUPAC Name1,1,1,2,3,3,3-heptafluoro-2-iodopropane
Molecular FormulaC3F7I
InChI KeyBBZVTTKMXRPMHZ-UHFFFAOYSA-N
SMILESFC(F)(F)C(F)(I)C(F)(F)F
さらに表示
Appearance (Color)Clear colorless to pink to purple
Assay (GC)≥96.0%
FormLiquid
Refractive Index1.3270-1.3390 @ 20°C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only