Pentamethyldisiloxane, 95%
Pentamethyldisiloxane, 95%
Pentamethyldisiloxane, 95%
Thermo Scientific Chemicals

Pentamethyldisiloxane, 95%

CAS: 1438-82-0 | C5H15OSi2 | 147.344 g/mol
製品番号(カタログ番号) L16577.14
または、製品番号L16577-14
価格(JPY)
-
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS1438-82-0
IUPAC Namedimethyl[(trimethylsilyl)oxy]silyl
Molecular FormulaC5H15OSi2
InChI KeyXUKFPAQLGOOCNJ-UHFFFAOYSA-N
SMILESC[Si](C)O[Si](C)(C)C
さらに表示
Appearance (Color)Clear colorless
FormLiquid
Refractive Index1.3710-1.3770 @ 20?C
Assay (GC)≥94.0%
Pentamethyldisiloxane is used in the synthesis of siloxane-polyether copolymer surfactants. It is also used as source of trimethylsilyl functional group in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pentamethyldisiloxane is used in the synthesis of siloxane-polyether copolymer surfactants. It is also used as source of trimethylsilyl functional group in organic synthesis.

Solubility
Not miscible or difficult to mix in water.

Notes
It is sensitive to moisture. Keep container tightly closed in a dry and well-ventilated place. Store under dry inert gas. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Tao Pei.; Ross A. Widenhoefer. Use of Pentamethyldisiloxane in the Palladium-Catalyzed Cyclization/Hydrosilylation of Functionalized Dienes. Org. Lett. 2000, 2 (10), 1469-1471.
  2. Carles, S.; Le Garrec, J. L.; Mitchell, J. B. Electron and ion reactions with hexamethyldisiloxane and pentamethyldisiloxane. The Journal of chemical physics. 2007, 127 (4), 144308-144308.