1H,1H-Perfluoro-1-decanol, 98%
1H,1H-Perfluoro-1-decanol, 98%
1H,1H-Perfluoro-1-decanol, 98%
Thermo Scientific Chemicals

1H,1H-Perfluoro-1-decanol, 98%

CAS: 307-37-9 | C10H3F19O | 500.103 g/mol
製品番号(カタログ番号) L16580.14
または、製品番号L16580-14
価格(JPY)
-
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS307-37-9
IUPAC Name2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-nonadecafluorodecan-1-ol
Molecular FormulaC10H3F19O
InChI KeyNIRPXSQCRWXHNZ-UHFFFAOYSA-N
SMILESOCC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F
さらに表示
Melting Point (clear melt)82-90°C
Appearance (Color)White or colorless to pale cream
Assay (GC)≥97.5%
FormPowder or crystalline powder or crystals
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only