Ishikawa's Reagent, 90+%
Ishikawa's Reagent, 90+%
Ishikawa's Reagent, 90+%
Thermo Scientific Chemicals

Ishikawa's Reagent, 90+%

CAS: 309-88-6 | C7H11F6N | 223.162 g/mol
製品番号(カタログ番号) L16738.14
または、製品番号L16738-14
価格(JPY)
-
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS309-88-6
IUPAC Namediethyl(1,1,2,3,3,3-hexafluoropropyl)amine
Molecular FormulaC7H11F6N
InChI KeyBNTFCVMJHBNJAR-UHFFFAOYNA-N
SMILESCCN(CC)C(F)(F)C(F)C(F)(F)F
さらに表示
FormLiquid
CommentMaterial Sourced in the U.S. and in other countries
Appearance (Color)Clear colorless to yellow
Assay (GC)≥90.0%
Refractive Index1.3145-1.3945 @ 20°C (non-U.S. sourced material)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only