4,4,4-Trifluoro-3-hydroxy-3-methylbutyric acid, 98%
4,4,4-Trifluoro-3-hydroxy-3-methylbutyric acid, 98%
4,4,4-Trifluoro-3-hydroxy-3-methylbutyric acid, 98%
Thermo Scientific Chemicals

4,4,4-Trifluoro-3-hydroxy-3-methylbutyric acid, 98%

CAS: 338-03-4 | C5H7F3O3 | 172.103 g/mol
製品番号(カタログ番号) L16873.06
または、製品番号L16873-06
価格(JPY)
-
数量:
5 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS338-03-4
IUPAC Name(3S)-4,4,4-trifluoro-3-hydroxy-3-methylbutanoate
Molecular FormulaC5H6F3O3
InChI KeyJCJGOWYCFRASTM-BYPYZUCNSA-M
SMILESC[C@](O)(CC([O-])=O)C(F)(F)F
さらに表示
Appearance (Color)White or clear colorless to pale yellow
FormFused solid or liquid as melt
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
Assay (Silylated GC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only