Glycine tert-butyl ester hydrochloride, 99%
Glycine tert-butyl ester hydrochloride, 99%
Glycine tert-butyl ester hydrochloride, 99%
Thermo Scientific Chemicals

Glycine tert-butyl ester hydrochloride, 99%

CAS: 27532-96-3 | C6H14NO2 | 132.18 g/mol
製品番号(カタログ番号) L17116.03
または、製品番号L17116-03
価格(JPY)
-
数量:
1 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS27532-96-3
IUPAC Nametert-butyl 2-aminoacetate hydrochloride
Molecular FormulaC6H14ClNO2
InChI KeyOSWULUXZFOQIRU-UHFFFAOYSA-N
SMILESCl.CC(C)(C)OC(=O)CN
さらに表示
Assay (Freebase GC)≥96.0%
Appearance (Color)White to pale cream
Assay (Titration ex Chloride)≥96.0 to ≤104.0%
FormCrystals or powder or crystalline powder or lumps
Identification (FTIR)Conforms
Glycine t-butyl ester hydrochloride is a substituted Glycine used as parakeratosis inhibitor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Glycine t-butyl ester hydrochloride is a substituted Glycine used as parakeratosis inhibitor.

Solubility
Soluble in methanol and water

Notes
Keep container tightly closed. Store away from oxidizing agents.
RUO – Research Use Only

General References:

  1. H. R. Allcock.; T. J. Fuller.; D. P. Mack.; K. Matsumura.; Karen M. Smeltz. Synthesis of Poly[(amino acid alkyl ester)phosphazenes]. Macromolecules. 1977, 10 (4), 824-830.
  2. Hiroyuki Ishiwata.; Hiroki Sone.; Hideo Kigoshi.; Kiyoyuki Yamada. Total Synthesis of Doliculide, a Potent Cytotoxic Cyclodepsipeptide from the Japanese Sea Hare Dolabella auricularia. J. Org. Chem. 1994, 59 (17), 4712-4713.