Thermo Scientific Chemicals

6-Cyanonicotinic acid, 97%, Thermo Scientific Chemicals

Catalog number: L17127.MD
250 mg, Each
Thermo Scientific Chemicals

6-Cyanonicotinic acid, 97%, Thermo Scientific Chemicals

Catalog number: L17127.MD
250 mg, Each
Quantity
Catalog number: L17127.MD
also known as L17127-MD
Price (USD)
Price: 94.90
Online price: 81.65
Your price:
Quantity
-

Chemical Identifiers

CAS
70165-31-0
IUPAC Name
6-cyanopyridine-3-carboxylic acid
Molecular Formula
C7H4N2O2
InChI Key
WMHSQCDPPJRWIL-UHFFFAOYSA-N
SMILES
OC(=O)C1=CN=C(C=C1)C#N
Appearance (Color)
Pale yellow to orange or brown
Form
Crystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)
≥96.0% to ≤104.0%
Assay (HPLC)
≥96.0%
Melting Point (clear melt)
187-195°C

Description

6-Cyanonicotinic acid, is used in medicine

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Cyanonicotinic acid, is used in medicine

Solubility
Soluble in alcohol.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text