5-Bromo-2-iodopyrimidine, 98%, Thermo Scientific Chemicals
5-Bromo-2-iodopyrimidine, 98%, Thermo Scientific Chemicals
5-Bromo-2-iodopyrimidine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

5-Bromo-2-iodopyrimidine, 98%, Thermo Scientific Chemicals

製品番号(カタログ番号)数量
L17453.06
または、製品番号L17453-06
5 g
製品番号(カタログ番号) L17453.06
または、製品番号L17453-06
価格(JPY)
-
数量:
5 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS183438-24-6
IUPAC Name5-bromo-2-iodopyrimidine
Molecular FormulaC4H2BrIN2
InChI KeyZEZKXPQIDURFKA-UHFFFAOYSA-N
SMILESBrC1=CN=C(I)N=C1
さらに表示
Appearance (Color)White to cream to yellow to orange to brown to grey to dark grey
FormCrystals or powder or crystalline powder or fused solid or flakes
Assay (GC)≥97.5%
Identification (FTIR)Conforms
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only