3,4-Dimethylbenzeneboronic acid, 98+%
3,4-Dimethylbenzeneboronic acid, 98+%
3,4-Dimethylbenzeneboronic acid, 98+%
Thermo Scientific Chemicals

3,4-Dimethylbenzeneboronic acid, 98+%

CAS: 55499-43-9 | C8H11BO2 | 149.98 g/mol
製品番号(カタログ番号) L17461.03
または、製品番号L17461-03
価格(JPY)
-
数量:
1 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS55499-43-9
IUPAC Name(3,4-dimethylphenyl)boronic acid
Molecular FormulaC8H11BO2
InChI KeyKDVZJKOYSOFXRV-UHFFFAOYSA-N
SMILESCC1=CC=C(C=C1C)B(O)O
さらに表示
Assay (HPLC)≥98.0%
Proton NMRConforms to structure
Appearance (Color)White to pale cream
FormPowder
Assay (Aqueous acid-base Titration)≥98.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only